Assay Details
Gene Symbol: Ccar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026201.3 | NP_080477.1 | 1 - 2 | 149 | 104 | |
GenBank mRNA | AF465615.1 | - | 1 - 2 | 40 | 104 | |
AK012111.1 | - | 1 - 2 | 40 | 104 | ||
BC034174.1 | - | 1 - 2 | 50 | 104 | ||
BC039939.1 | - | 1 - 2 | 49 | 105 | ||
BC051052.1 | - | 1 - 2 | 51 | 104 | ||
BC060130.1 | - | 1 - 2 | 37 | 104 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF465615.1, AK012111.1, BC034174.1, BC039939.1, BC051052.1, BC060130.1, NM_026201.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
