Assay Details
Gene Symbol: Snx14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172926.3 | NP_766514.2 | 20 - 21 | 2223 | 158 | |
GenBank mRNA | AK049371.1 | - | 20 - 21 | 2202 | 158 | |
AK082389.1 | - | 20 - 21 | 2144 | 158 | ||
BC043328.1 | - | 5 - 6 | 881 | 158 | ||
BC080799.1 | - | 20 - 21 | 2208 | 4948538 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
