Assay Details
Gene Symbol: Sltm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025690.3 | NP_079966.2 | 18 - 19 | 2730 | 84 | |
NM_026337.1 | NP_080613.1 | 18 - 19 | 2768 | 84 | ||
GenBank mRNA | AF462146.1 | - | 18 - 19 | 2730 | 84 | |
BC019992.1 | - | 7 - 8 | 1100 | 84 | ||
BC139202.1 | - | 18 - 19 | 2720 | 8862224 | 84 | |
BC145577.1 | - | 17 - 18 | 2539 | 84 | ||
CF162411.1 | - | 2 - 3 | 326 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
