Assay Details
Gene Symbol: Sltm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025690.3 | NP_079966.2 | 8 - 9 | 1161 | 110 | |
NM_026337.1 | NP_080613.1 | 8 - 9 | 1199 | 110 | ||
GenBank mRNA | AF462146.1 | - | 8 - 9 | 1161 | 110 | |
AK011815.1 | - | 8 - 9 | 1147 | 110 | ||
AK017715.1 | - | 5 - 6 | 839 | 110 | ||
AK017839.1 | - | 8 - 9 | 1060 | 110 | ||
AK045723.1 | - | 2 - 3 | 261 | 110 | ||
AK077578.1 | - | 8 - 9 | 1222 | 110 | ||
AK082294.1 | - | 5 - 6 | 2077 | 110 | ||
BC139202.1 | - | 8 - 9 | 1151 | 8862224 | 110 | |
BC145577.1 | - | 8 - 9 | 1151 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
