Assay Details
Gene Symbol: Ipmk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027184.1 | NP_081460.1 | 2 - 3 | 388 | 62 | |
GenBank mRNA | AK010523.1 | - | 2 - 3 | 347 | 62 | |
AK036084.1 | - | 2 - 3 | 156 | 62 | ||
AK036978.1 | - | 2 - 3 | 348 | 62 | ||
AK172246.1 | - | 2 - 3 | 346 | 62 | ||
BC052463.1 | - | 2 - 3 | 388 | 6834748 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
