Assay Details
Gene Symbol: Tjp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282095.1 | NP_001269024.1 | 7 - 8 | 986 | 61 | |
NM_001282096.1 | NP_001269025.1 | 7 - 8 | 1049 | 61 | ||
NM_013769.3 | NP_038797.2 | 7 - 8 | 1088 | 61 | ||
GenBank mRNA | AF157006.1 | - | 7 - 8 | 1056 | 61 | |
AK029083.1 | - | 7 - 8 | 1035 | 61 | ||
AK134100.1 | - | 7 - 8 | 1089 | 61 | ||
AK167587.1 | - | 7 - 8 | 987 | 61 | ||
BC012518.1 | - | 7 - 8 | 1053 | 3600848 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
