Assay Details
Gene Symbol: Sirt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122765.1 | NP_001116237.1 | 13 - 14 | 1002 | 76 | |
NM_001122766.1 | NP_001116238.1 | 11 - 12 | 839 | 76 | ||
NM_022432.4 | NP_071877.3 | 14 - 15 | 1049 | 76 | ||
GenBank mRNA | AB221570.1 | - | 14 - 15 | 954 | 76 | |
AF299337.1 | - | 14 - 15 | 1034 | 76 | ||
AK014042.1 | - | 13 - 14 | 973 | 76 | ||
AK144431.1 | - | 13 - 14 | 946 | 76 | ||
AK146412.1 | - | 13 - 14 | 979 | 76 | ||
BC021439.1 | - | 14 - 15 | 1053 | 5039278 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
