Assay Details
Gene Symbol: Adarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024837.2 | NP_001020008.1 | 7 - 8 | 1816 | 65 | |
NR_004429.1 | 7 - 8 | 1863 | 65 | |||
GenBank mRNA | AF403107.2 | - | 7 - 8 | 1768 | 65 | |
AF403109.2 | - | 7 - 8 | 1815 | 65 | ||
AK141777.1 | - | 7 - 8 | 1788 | 65 | ||
AK147298.1 | - | 8 - 9 | 1895 | 65 | ||
BC040200.1 | - | 3 - 4 | 371 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
