Assay Details
Gene Symbol: Pfkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008826.4 | NP_032852.2 | 12 - 13 | 1299 | 100 | |
GenBank mRNA | AK030511.1 | - | 12 - 13 | 1254 | 100 | |
AK030892.1 | - | 5 - 6 | 398 | 100 | ||
AK036318.1 | - | 12 - 13 | 1232 | 100 | ||
AK081313.1 | - | 12 - 13 | 1250 | 100 | ||
AK159328.1 | - | 12 - 13 | 1244 | 100 | ||
AK168660.1 | - | 12 - 13 | 1255 | 100 | ||
AK179375.1 | - | 2 - 3 | 116 | 100 | ||
AK184815.1 | - | 2 - 3 | 116 | 100 | ||
AK209868.1 | - | 2 - 3 | 116 | 100 | ||
BC020097.1 | - | 12 - 13 | 1253 | 4014395 | 100 | |
BC029336.1 | - | 12 - 13 | 1243 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
