Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900021323.mm.1 |
TC0900002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021323.mm.1 |
NM_001164248.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021323.mm.1 |
NM_001164248.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021323.mm.1 |
NM_001164251.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021323.mm.1 |
NM_001164251.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021323.mm.1 |
NM_001164254.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021323.mm.1 |
NM_001164254.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021322.mm.1 |
TC0900002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021322.mm.1 |
NM_001164248.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021322.mm.1 |
NM_001164248.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021322.mm.1 |
NM_001164251.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021322.mm.1 |
NM_001164251.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021322.mm.1 |
NM_001164254.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021322.mm.1 |
NM_001164254.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021320.mm.1 |
TC0900002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021320.mm.1 |
NM_001164248.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021320.mm.1 |
NM_001164248.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021320.mm.1 |
NM_001164251.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021320.mm.1 |
NM_001164251.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021320.mm.1 |
NM_001164254.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021320.mm.1 |
NM_001164254.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021327.mm.1 |
TC0900002541.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900021327.mm.1 |
NM_001164248.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021327.mm.1 |
NM_001164248.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021327.mm.1 |
NM_001164251.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021327.mm.1 |
NM_001164251.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021327.mm.1 |
NM_001164254.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900021327.mm.1 |
NM_001164254.1 |
Refseq |