Assay Details
Gene Symbol: Grik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010348.3 | NP_034478.1 | 8 - 9 | 1765 | 96 | |
NM_146072.4 | NP_666184.2 | 8 - 9 | 1765 | 96 | ||
GenBank mRNA | AK042635.1 | - | 8 - 9 | 1766 | 96 | |
AK043542.1 | - | 8 - 9 | 1745 | 96 | ||
AK043737.1 | - | 8 - 9 | 1729 | 96 | ||
AK044250.1 | - | 8 - 9 | 1724 | 96 | ||
AK044420.1 | - | 8 - 9 | 1720 | 96 | ||
AK048634.1 | - | 8 - 9 | 1736 | 96 | ||
AK133667.1 | - | 8 - 9 | 1751 | 96 | ||
AK200858.1 | - | 2 - 3 | 260 | 96 | ||
BC031822.1 | - | 7 - 8 | 1187 | 4501004 | 96 | |
BC049275.1 | - | 7 - 8 | 1187 | 4501004 | 96 | |
X66118.1 | - | 8 - 9 | 1751 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
