Assay Details
Gene Symbol: Slc44a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199186.1 | NP_001186115.1 | 13 - 14 | 1243 | 88 | |
NM_152808.3 | NP_690021.1 | 13 - 14 | 1245 | 88 | ||
GenBank mRNA | AK036249.1 | - | 1 - 2 | 689 | 88 | |
AK041533.1 | - | 13 - 14 | 1202 | 88 | ||
AK048648.1 | - | 13 - 14 | 1230 | 88 | ||
AK153992.1 | - | 13 - 14 | 1224 | 88 | ||
AK166695.1 | - | 13 - 14 | 1246 | 88 | ||
BC013213.1 | - | 6 - 7 | 638 | 88 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
