Assay Details
Gene Symbol: Taf1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028401.1 | 11 - 12 | 1184 | 113 | ||
GenBank mRNA | AK012656.1 | - | 9 - 10 | 1033 | 113 | |
AK014920.1 | - | 11 - 12 | 1136 | 113 | ||
AK050165.1 | - | 11 - 12 | 1140 | 113 | ||
AK166329.1 | - | 11 - 12 | 1139 | 113 | ||
AK166633.1 | - | 10 - 11 | 2254 | 113 | ||
BC110660.1 | - | 8 - 9 | 2161 | 3158156 | 113 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
