Assay Details
Gene Symbol: Prkcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293650.1 | NP_001280579.1 | 15 - 16 | 1499 | 91 | |
NM_001293651.1 | NP_001280580.1 | 15 - 16 | 1482 | 91 | ||
NM_008925.2 | NP_032951.1 | 15 - 16 | 1478 | 91 | ||
GenBank mRNA | AK137661.1 | - | 15 - 16 | 1445 | 91 | |
AK153931.1 | - | 15 - 16 | 1453 | 91 | ||
AK159705.1 | - | 15 - 16 | 1449 | 91 | ||
BC009816.1 | - | 15 - 16 | 1432 | 4008182 | 91 | |
U92794.1 | - | 15 - 16 | 1431 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
