Assay Details
Gene Symbol: Arhgap42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027823.1 | NP_082099.1 | 4 - 5 | 622 | 77 | |
GenBank mRNA | AK082857.1 | - | 4 - 5 | 711 | 77 | |
BC137891.1 | - | 4 - 5 | 622 | 8860910 | 77 | |
BC137892.1 | - | 4 - 5 | 622 | 8860911 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
