Assay Details
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011680.2 | NP_035810.1 | 2 - 3 | 153 | 90 | |
GenBank mRNA | AB221562.1 | - | 2 - 3 | 112 | 90 | |
AK146871.1 | - | 2 - 3 | 100 | 90 | ||
BC082995.1 | - | 2 - 3 | 153 | 6440967 | 90 | |
U01662.1 | - | 2 - 3 | 169 | 90 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
