Assay Details
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011680.2 | NP_035810.1 | 6 - 7 | 715 | 60 | |
GenBank mRNA | AB221562.1 | - | 6 - 7 | 674 | 60 | |
AK146871.1 | - | 6 - 7 | 662 | 60 | ||
AK158906.1 | - | 4 - 5 | 491 | 60 | ||
BC019729.1 | - | 1 - 2 | 95 | 60 | ||
BC082995.1 | - | 6 - 7 | 715 | 6440967 | 60 | |
U01662.1 | - | 6 - 7 | 731 | 60 | ||
U01663.1 | - | 3 - 4 | 268 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
