Assay Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081346.1 | NP_001074815.1 | 1 - 2 | 218 | 96 | |
GenBank mRNA | AK045134.1 | - | 119 | 96 | ||
AK052662.1 | - | 1 - 2 | 247 | 96 | ||
BC116341.2 | - | 211 | 40098130 | 96 | ||
BC116342.2 | - | 211 | 40098131 | 96 | ||
BC158093.1 | - | 1 - 2 | 214 | 9007380 | 96 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK045134.1, AK052662.1, BC116341.2, BC116342.2, BC158093.1, NM_001081346.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
