Assay Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081346.1 | NP_001074815.1 | 5 - 6 | 647 | 106 | |
GenBank mRNA | AK030986.1 | - | 5 - 6 | 531 | 106 | |
AK045134.1 | - | 548 | 106 | |||
AK052662.1 | - | 5 - 6 | 685 | 106 | ||
BC116341.2 | - | 649 | 40098130 | 106 | ||
BC116342.2 | - | 649 | 40098131 | 106 | ||
BC158093.1 | - | 5 - 6 | 652 | 9007380 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
