Assay Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081346.1 | NP_001074815.1 | 9 - 10 | 1043 | 67 | |
GenBank mRNA | AK030986.1 | - | 8 - 9 | 924 | 67 | |
AK045134.1 | - | 941 | 67 | |||
AK052662.1 | - | 8 - 9 | 1078 | 67 | ||
AK082859.1 | - | 1 - 2 | 81 | 67 | ||
BC116341.2 | - | 1042 | 40098130 | 67 | ||
BC116342.2 | - | 1042 | 40098131 | 67 | ||
BC158093.1 | - | 8 - 9 | 1045 | 9007380 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
