Assay Details
Gene Symbol: Tbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019786.4 | NP_062760.3 | 17 - 18 | 2018 | 115 | |
GenBank mRNA | AF145705.1 | - | 16 - 17 | 1870 | 115 | |
AF191839.1 | - | 16 - 17 | 1897 | 115 | ||
AK004269.1 | - | 1 - 2 | 80 | 115 | ||
AK083214.1 | - | 17 - 18 | 1966 | 115 | ||
BC129910.1 | - | 16 - 17 | 1930 | 40111752 | 115 | |
BC129911.1 | - | 16 - 17 | 1930 | 40111754 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
