Assay Details
Gene Symbol: Elp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021448.2 | NP_067423.2 | 14 - 15 | 1574 | 84 | |
GenBank mRNA | AF291064.1 | - | 14 - 15 | 1540 | 84 | |
AK033246.1 | - | 14 - 15 | 1575 | 84 | ||
AK034457.1 | - | 14 - 15 | 1573 | 84 | ||
AK050960.1 | - | 14 - 15 | 1575 | 84 | ||
AK147826.1 | - | 1575 | 84 | |||
AK167725.1 | - | 11 - 12 | 1303 | 84 | ||
AK167853.1 | - | 14 - 15 | 1573 | 84 | ||
AK168370.1 | - | 14 - 15 | 1575 | 84 | ||
BC015284.1 | - | 14 - 15 | 1566 | 4239056 | 84 | |
BC051943.1 | - | 14 - 15 | 1566 | 5686798 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
