Assay Details
Gene Symbol: Strip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037740.1 | NP_001032829.1 | 10 - 11 | 1364 | 96 | |
NM_177204.3 | NP_796178.2 | 10 - 11 | 1364 | 96 | ||
GenBank mRNA | AK041004.1 | - | 10 - 11 | 1365 | 96 | |
AK042073.1 | - | 10 - 11 | 1365 | 96 | ||
AK052273.1 | - | 2 - 3 | 389 | 96 | ||
AK122459.1 | - | 10 - 11 | 1169 | 96 | ||
AK163706.1 | - | 10 - 11 | 1364 | 96 | ||
BC147502.1 | - | 10 - 11 | 1356 | 9056385 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
