Assay Details
Gene Symbol: Pik3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181414.5 | NP_852079.2 | 17 - 18 | 2024 | 153 | |
GenBank mRNA | AK030578.1 | - | 17 - 18 | 2054 | 153 | |
AK044997.1 | - | 17 - 18 | 2023 | 153 | ||
BC027029.1 | - | 17 - 18 | 2066 | 153 | ||
BC057678.1 | - | 17 - 18 | 2027 | 3496633 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
