Assay Details
Gene Symbol: Vwa3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177697.3 | NP_808365.2 | 13 - 14 | 1638 | 67 | |
GenBank mRNA | AK086942.1 | - | 12 - 13 | 1236 | 67 | |
AK132732.1 | - | 13 - 14 | 1639 | 67 | ||
AK136898.1 | - | 12 - 13 | 1237 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
