Assay Details
Gene Symbol: Map3k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011946.3 | NP_036076.2 | 3 - 4 | 654 | 93 | |
GenBank mRNA | AK143367.1 | - | 3 - 4 | 651 | 93 | |
U43186.1 | - | 3 - 4 | 551 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
