Assay Details
Gene Symbol: Map3k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011946.3 | NP_036076.2 | 9 - 10 | 1204 | 65 | |
GenBank mRNA | U43186.1 | - | 9 - 10 | 1102 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
