Assay Details
Gene Symbol: Iws1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173441.3 | NP_775617.1 | 8 - 9 | 1917 | 73 | |
GenBank mRNA | AK006925.1 | - | 2 - 3 | 210 | 73 | |
AK028247.1 | - | 8 - 9 | 1888 | 73 | ||
AK033013.1 | - | 8 - 9 | 1912 | 73 | ||
AK136492.1 | - | 4 - 5 | 373 | 73 | ||
BC068184.1 | - | 7 - 8 | 2503 | 30639737 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
