Assay Details
Gene Symbol: Ercc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133658.1 | NP_598419.1 | 10 - 11 | 1796 | 68 | |
GenBank mRNA | AK151188.1 | - | 10 - 11 | 1801 | 68 | |
AK152347.1 | - | 10 - 11 | 1801 | 68 | ||
AK152988.1 | - | 10 - 11 | 1808 | 68 | ||
AK154745.1 | - | 10 - 11 | 1802 | 68 | ||
AK160187.1 | - | 4 - 5 | 900 | 68 | ||
BC016595.1 | - | 10 - 11 | 1767 | 3153847 | 68 | |
BC022956.1 | - | 9 - 10 | 1900 | 68 | ||
BC024446.1 | - | 1 - 2 | 142 | 68 | ||
BC026575.1 | - | 10 - 11 | 1778 | 4955015 | 68 | |
S71186.1 | - | 10 - 11 | 1796 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
