Assay Details
Gene Symbol: Ercc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133658.1 | NP_598419.1 | 11 - 12 | 1894 | 68 | |
GenBank mRNA | AK151188.1 | - | 11 - 12 | 1899 | 68 | |
AK152347.1 | - | 11 - 12 | 1899 | 68 | ||
AK152988.1 | - | 11 - 12 | 1906 | 68 | ||
AK154745.1 | - | 11 - 12 | 1900 | 68 | ||
AK160187.1 | - | 5 - 6 | 998 | 68 | ||
BC016595.1 | - | 11 - 12 | 1865 | 3153847 | 68 | |
BC022956.1 | - | 10 - 11 | 1998 | 68 | ||
BC024446.1 | - | 2 - 3 | 240 | 68 | ||
BC026575.1 | - | 11 - 12 | 1876 | 4955015 | 68 | |
S71186.1 | - | 11 - 12 | 1894 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
