Assay Details
Gene Symbol: Ercc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133658.1 | NP_598419.1 | 6 - 7 | 882 | 78 | |
GenBank mRNA | AK151188.1 | - | 6 - 7 | 887 | 78 | |
AK152347.1 | - | 6 - 7 | 887 | 78 | ||
AK152988.1 | - | 6 - 7 | 889 | 79 | ||
AK154745.1 | - | 6 - 7 | 888 | 78 | ||
BC016595.1 | - | 6 - 7 | 853 | 3153847 | 78 | |
BC022956.1 | - | 5 - 6 | 986 | 78 | ||
BC026575.1 | - | 6 - 7 | 864 | 4955015 | 78 | |
S71186.1 | - | 6 - 7 | 882 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
