Assay Details
Gene Symbol: Arf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007480.1 | NP_031506.1 | 3 - 4 | 306 | 59 | |
GenBank mRNA | BC132452.1 | - | 4 - 5 | 294 | 40130729 | 59 |
BC138756.1 | - | 4 - 5 | 294 | 8861778 | 59 | |
D87902.1 | - | 3 - 4 | 306 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
