Assay Details
Gene Symbol: Phtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172992.3 | NP_766580.3 | 6 - 7 | 699 | 59 | |
GenBank mRNA | AK035536.1 | - | 6 - 7 | 653 | 59 | |
AK037021.1 | - | 6 - 7 | 586 | 59 | ||
AK042382.1 | - | 6 - 7 | 678 | 59 | ||
AK152951.1 | - | 6 - 7 | 652 | 59 | ||
BC052841.1 | - | 6 - 7 | 612 | 30050876 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
