Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000011152.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011152.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011152.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011152.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011152.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011152.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011152.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011148.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011148.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011148.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011148.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011148.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011148.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011148.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011151.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011151.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011151.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011151.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011151.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011151.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011151.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011138.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011138.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011138.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011138.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011138.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011137.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011137.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011137.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011137.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011137.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011162.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011162.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011162.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011162.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011162.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011162.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011162.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011139.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011139.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011139.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011139.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011139.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011150.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011150.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011150.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011150.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011150.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011150.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011150.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011153.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011153.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011153.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011153.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011153.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011153.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011153.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011160.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011160.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011160.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011157.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011157.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011157.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011157.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011157.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011157.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011157.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011158.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011158.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011158.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011158.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011158.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011158.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011158.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011161.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011161.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011161.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011161.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011161.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011161.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011161.mm.1 |
NM_027900.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011165.mm.1 |
TC1000001561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000011165.mm.1 |
NM_001168292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011165.mm.1 |
NM_001168292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011165.mm.1 |
NM_001168293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011165.mm.1 |
NM_001168293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011165.mm.1 |
NM_027900.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000011165.mm.1 |
NM_027900.4 |
Refseq |