Assay Details
Gene Symbol: Spon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145584.2 | NP_663559.1 | 12 - 13 | 1996 | 69 | |
GenBank mRNA | AK146506.1 | - | 12 - 13 | 1820 | 69 | |
BC020531.1 | - | 12 - 13 | 1866 | 4221758 | 69 | |
BC030339.1 | - | 3 - 4 | 450 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
