Assay Details
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023580.4 | NP_076069.2 | 7 - 8 | 1525 | 77 | |
GenBank mRNA | AF131197.1 | - | 7 - 8 | 1467 | 77 | |
AK028478.1 | - | 7 - 8 | 1526 | 77 | ||
BC071215.1 | - | 7 - 8 | 1489 | 4196138 | 77 | |
U18084.1 | - | 3 - 4 | 628 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
