Assay Details
Gene Symbol: Cdc25c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK141737.1 | - | 2 - 3 | 314 | 82 | |
AK168287.1 | - | 2 - 3 | 309 | 82 | ||
U15562.1 | - | 2 - 3 | 280 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK141737.1, AK168287.1, U15562.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
