Assay Details
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027994.1 | NP_082270.1 | 10 - 11 | 3278 | 108 | |
GenBank mRNA | AK078117.1 | - | 1 - 2 | 505 | 108 | |
AK129225.1 | - | 10 - 11 | 3239 | 108 | ||
BC057457.1 | - | 3 - 4 | 1921 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
