Assay Details
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027994.1 | NP_082270.1 | 13 - 14 | 3702 | 126 | |
GenBank mRNA | AK009683.1 | - | 2 - 3 | 341 | 126 | |
AK078117.1 | - | 4 - 5 | 929 | 126 | ||
AK129225.1 | - | 13 - 14 | 3663 | 126 | ||
BC057457.1 | - | 6 - 7 | 2345 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
