Assay Details
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027994.1 | NP_082270.1 | 9 - 10 | 1778 | 80 | |
GenBank mRNA | AK129225.1 | - | 9 - 10 | 1739 | 80 | |
AK144869.1 | - | 9 - 10 | 1770 | 80 | ||
AK158080.1 | - | 9 - 10 | 1774 | 80 | ||
BC057457.1 | - | 2 - 3 | 421 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
