Assay Details
Gene Symbol: Eps8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133191.2 | NP_573454.2 | 9 - 10 | 873 | 66 | |
GenBank mRNA | BC005492.1 | - | 9 - 10 | 873 | 3487337 | 66 |
BC009098.1 | - | 9 - 10 | 931 | 3482190 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
