Assay Details
Gene Symbol: B4galnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177897.3 | NP_808565.2 | 8 - 9 | 789 | 87 | |
GenBank mRNA | AB114827.1 | - | 8 - 9 | 789 | 87 | |
BC031982.1 | - | 7 - 8 | 643 | 87 | ||
BC062653.1 | - | 4 - 5 | 479 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
