Assay Details
Gene Symbol: Mgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171003.1 | NP_001164474.1 | 39 - 40 | 4735 | 66 | |
GenBank mRNA | AK031332.1 | - | 23 - 24 | 2739 | 66 | |
BC025509.1 | - | 5 - 6 | 435 | 66 | ||
EU937529.1 | - | 39 - 40 | 4735 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
