Assay Details
Gene Symbol: Gls2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033264.3 | NP_001028436.2 | 7 - 8 | 927 | 157 | |
NM_001285777.1 | NP_001272706.1 | 6 - 7 | 847 | 157 | ||
NM_001285779.1 | NP_001272708.1 | 7 - 8 | 956 | 157 | ||
GenBank mRNA | AK039618.1 | - | 6 - 7 | 735 | 157 | |
AK165896.1 | - | 7 - 8 | 873 | 157 | ||
AK220231.1 | - | 7 - 8 | 879 | 157 | ||
BC141115.1 | - | 7 - 8 | 927 | 9055727 | 157 | |
EU770627.1 | - | 7 - 8 | 865 | 157 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001285779.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
