Assay Details
Gene Symbol: Inpp5f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178641.5 | NP_848756.2 | 17 - 18 | 2253 | 69 | |
GenBank mRNA | AK030870.1 | - | 17 - 18 | 2255 | 69 | |
AK034482.1 | - | 2 - 3 | 139 | 69 | ||
AK047166.1 | - | 3 - 4 | 184 | 69 | ||
AK129249.1 | - | 17 - 18 | 2210 | 69 | ||
AK154996.1 | - | 17 - 18 | 2258 | 69 | ||
BC050127.1 | - | 5 - 6 | 614 | 69 | ||
BC067200.1 | - | 3 - 4 | 374 | 69 | ||
BC125437.1 | - | 17 - 18 | 2141 | 40129952 | 69 | |
BC137700.1 | - | 17 - 18 | 2141 | 8860719 | 69 | |
DQ648020.1 | - | 3 - 4 | 896 | 69 | ||
DQ792944.1 | - | 3 - 4 | 189 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK034482.1, AK047166.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
