Assay Details
Gene Symbol: Rpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026632.4 | NP_080908.1 | 2 - 3 | 221 | 134 | |
GenBank mRNA | AK005285.1 | - | 2 - 3 | 194 | 134 | |
AK009916.1 | - | 2 - 3 | 193 | 134 | ||
AK021228.1 | - | 2 - 3 | 194 | 134 | ||
BC028489.1 | - | 2 - 3 | 186 | 1448430 | 134 | |
BY057686.1 | - | 2 - 3 | 222 | 134 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
