Assay Details
Gene Symbol: Copg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017478.3 | NP_059506.1 | 581 | 116 | ||
GenBank mRNA | AF205065.1 | - | 484 | 116 | ||
AJ251067.1 | - | 553 | 116 | |||
AK079599.1 | - | 7 - 8 | 497 | 116 | ||
AK088492.1 | - | 565 | 116 | |||
AK147943.1 | - | 527 | 116 | |||
AK149462.1 | - | 7 - 8 | 518 | 116 | ||
AK163668.1 | - | 7 - 8 | 552 | 116 | ||
BC049178.1 | - | 547 | 6309945 | 116 | ||
BC145898.1 | - | 524 | 40131201 | 116 | ||
BC145900.1 | - | 524 | 40131203 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
