Assay Details
Gene Symbol: Cped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081351.1 | NP_001074820.1 | 2 - 3 | 1338 | 78 | |
GenBank mRNA | BC151018.1 | - | 2 - 3 | 853 | 9087931 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
