Assay Details
Gene Symbol: Glra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183427.4 | NP_906272.1 | 4 - 5 | 594 | 94 | |
GenBank mRNA | AK135134.1 | - | 4 - 5 | 738 | 94 | |
AK139335.1 | - | 4 - 5 | 1007 | 94 | ||
BC056342.1 | - | 4 - 5 | 594 | 5708257 | 94 | |
BC068987.1 | - | 4 - 5 | 594 | 5708257 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
