Assay Details
Gene Symbol: Glra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183427.4 | NP_906272.1 | 5 - 6 | 695 | 85 | |
GenBank mRNA | AK135134.1 | - | 5 - 6 | 839 | 85 | |
AK139335.1 | - | 5 - 6 | 1108 | 85 | ||
BC037046.1 | - | 1 - 2 | 62 | 85 | ||
BC056342.1 | - | 5 - 6 | 695 | 5708257 | 85 | |
BC068987.1 | - | 5 - 6 | 695 | 5708257 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
